Learn More
Medchemexpress LLC Dbco-peg1-nhs ester | 2228857-34-7 | 95.0% | C28H27N3O7 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY140271
Encompass_Preferred
DBCO-PEG1-NHS ester is a PEG-based PROTAC linker designed for the synthesis of PROTACs. This click chemistry reagent contains a DBCO group, enabling it to undergo strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing azide groups.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Reacts with molecules containing azide groups
Catalog No. 50-003-52578