Learn More
Broadpharm DBCO-PEG12 100mg
Supplier:  Broadpharm BP29921100MG
Encompass
DBCO-PEG12-amine is a PEG linker which contains DBCO and amine moieties. The DBCO group is commonly used in copper-free Click Chemistry reactions. The amine group is reactive with carboxylic acids/ activated NHS esters/ carbonyls (ketone| aldehyde) etc. The hydrophilic PEG spacer increases the water solubility of the compound. Reagent grade for research purpose.
Catalog No. NC2892689