Learn More
Medchemexpress LLC Dbco-peg2-acid | 2304558-25-4 | ≥95.0% | C26H28N2O6 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140266100MG
Encompass_Preferred
DBCO-PEG2-acid is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. It functions as a click chemistry reagent, incorporating a DBCO group capable of undergoing strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules that contain Azide groups. PROTACs themselves are composed of two distinct ligands connected by a linker, one binding to an E3 ubiquitin ligase and the other to the target protein, enabling the selective degradation of target proteins through the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker.
  • Utilized in the synthesis of PROTACs.
  • Click chemistry reagent.
  • Contains a DBCO group for strain-promoted alkyne-azide cycloaddition (SPAAC) with azide groups.
  • PROTACs degrade target proteins via the ubiquitin-proteasome system.
Catalog No. 50-003-57117