Learn More
Medchemexpress LLC DBCO-PEG2-acid | 2304558-25-4 | >=95.0% | C26H28N2O6 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14026650MG
Encompass_Preferred
DBCO-PEG2-acid is a PEG-based PROTAC linker designed for the synthesis of PROTACs. It functions as a click chemistry reagent, featuring a DBCO group that enables strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing azide groups. PROTACs themselves are composed of two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets a specific protein, allowing PROTACs to exploit the intracellular ubiquitin-proteasome system for selective protein degradation.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Click chemistry reagent
  • Features a DBCO group for SPAAC
  • Reacts with azide-containing molecules
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-57118