Learn More
Medchemexpress LLC DBCO-PEG2-NHS ester | 2585653-12-7 | MFCD34551891 | ≥98.0% | 561.58 | C30H31N3O8 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15182710MG
Encompass_Preferred
DBCO-PEG2-NHS ester is an amine-reactive, click-chemistry reagent that installs a dibenzocyclooctyne (DBCO) handle via an N-hydroxysuccinimide (NHS) ester and a short PEG2 spacer. It is designed for efficient bioconjugation and copper-free strain-promoted azide-alkyne cycloaddition in research applications.
  • Amine-reactive NHS ester enables rapid and selective coupling to primary amines.
  • DBCO functionality supports copper-free strain-promoted click chemistry.
  • P EG2 spacer improves solubility and reduces steric hindrance for conjugation.
  • High purity (≥98.0%) suitable for sensitive research workflows.
  • Stable as powder at -20°C for long-term storage.
Catalog No. 50-002-86544