Learn More
Medchemexpress LLC Dbco-peg2-val-cit-pab-mmae | 2845934-59-8 | MFCD35133151 | 99.3% | 1569.92 | C84H120N12O17 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1722875MG
Encompass_Preferred
Dbco-peg2-val-cit-pab-mmae is a DBCO-functional antibody-drug conjugate linker for strain-promoted alkyne-azide cycloaddition (SPAAC) conjugation and intracellular protease-mediated release of MMAE; intended for research use only.
  • Click chemistry DBCO handle for strain-promoted alkyne-azide cycloaddition.
  • PEG2 spacer to improve solubility and spacing.
  • Val-Cit protease-cleavable dipeptide enabling intracellular release of payload.
  • PAB self-immolative spacer to facilitate payload release.
  • Contains MMAE cytotoxic payload for ADC research applications.
  • High purity (~99.3%) with recommended storage at -20°C, protected from light and under nitrogen.
Catalog No. 50-002-03646