Learn More
Medchemexpress LLC DBCO-PEG24-acid | 2765066-36-0 | C70H116N2O28 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1518205MG
Encompass_Preferred
DBCO-PEG24-acid is a click chemistry reagent and an analog of DBCO-Acid featuring a PEG linker and a DBCO group. The DBCO group is utilized in copper-free Click Chemistry reactions due to its strain-promoted high energy. The hydrophilic PEG chain enhances water solubility. The terminal carboxylic acid can react with primary amine groups in the presence of activators (e.g., EDC or HATU) to form a stable amide bond. This product is reagent grade and intended for research use only.
  • Click chemistry reagent
  • Analog of DBCO-Acid with PEG linker and DBCO group
  • DBCO group for copper-free Click Chemistry reactions
  • Strain-promoted high energy DBCO group
  • Hydrophilic PEG chain enhances water solubility
  • Terminal carboxylic acid reacts with primary amine groups
  • Forms stable amide bond with activators (e.g., EDC or HATU)
  • Reagent grade
  • Intended for research use only
Catalog No. 50-003-75011