Learn More
Medchemexpress LLC DBCO-PEG24-maleimide | 2924872-84-2 | MFCD31560742 | 98.0% | 1555.81 g·mol⁻¹ | C76H122N4O29 | 10mg
SDP
Supplier:  Medchemexpress LLC HYW87904810MG
Encompass_Preferred
DBCO-PEG24-Maleimide contains a maleimide and a DBCO group DBCO-PEG24-Maleimide can be used for thiol-containing biomolecule conjugations DBCO-PEG24-Maleimide can be used in click chemistry with azide-bearing biomolecules[1]
Catalog No. 50-002-70465