Learn More
Medchemexpress LLC DBCO-PEG24-NHS ester | 2765066-38-2 | C74H119N3O30 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY151835
Encompass_Preferred
DBCO-PEG24-NHS ester is a click chemistry reagent that reacts specifically and efficiently with primary amines under neutral or slightly basic conditions to form a covalent bond. The hydrophilic PEG spacer arm improves water solubility and provides a long, flexible connection, minimizing steric hindrance during ligation. DBCO is commonly used for copper-free click chemistry reactions.
  • Contains an NHS ester for covalent bond formation
  • Enhances water solubility
  • Minimizes steric hindrance during ligation
  • Suitable for copper-free click chemistry reactions
Catalog No. 50-003-74358