Learn More
Medchemexpress LLC Dbco-peg24-nhs ester | 2765066-38-2 | C74H119N3O30 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1518355MG
Encompass_Preferred
DBCO-PEG24-NHS ester is a click chemistry reagent that reacts specifically and efficiently with primary amines at neutral or slightly basic conditions to form a covalent bond. DBCO is commonly used for copper-free Click Chemistry reactions. It is for research use only.
  • The hydrophilic PEG spacer arm improves water solubility
  • Provides a long and flexible connection that minimizes steric hindrance involved with ligation
Catalog No. 50-003-73949