Learn More
Medchemexpress LLC DBCO-PEG3-acid | 2754384-59-1 | 98.4% | 508.56 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY42974100MG
Encompass_Preferred
DBCO-PEG3-acid is a non-cleavable 3 unit PEG ADC linker used in the synthesis of antibody-drug conjugates (ADCs). DBCO-PEG3-acid contains a DBCO group that can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups.
  • Non-cleavable 3 unit PEG linker
  • Used in antibody-drug conjugates (ADCs) synthesis
  • Contains a DBCO group
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) with azide groups
Catalog No. 50-004-03899