Learn More
Medchemexpress LLC Dbco-PEG4-acetic-val-cit-pab | 96.7% | 855.97 | C45H57N7O10 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY13609825MG
Encompass_Preferred
DBCO-PEG4-acetic-Val-Cit-PAB is a cleavable four-unit PEG linker containing a dibenzocyclooctyne (DBCO) group and a Val-Cit-PAB cleavable spacer, designed for use in antibody-drug conjugate synthesis and click-chemistry conjugation.
  • Contains DBCO for strain-promoted alkyne-azide cycloaddition (SPAAC).
  • Val-Cit-PAB spacer is enzymatically cleavable for intracellular payload release.
  • Designed for assembly of antibody-drug conjugates and payload conjugation.
  • Available in small pack sizes suitable for research applications.
  • Powder stable at -20°C for long-term storage; solution stability is temperature dependent.
Catalog No. 50-002-04169