Learn More
Medchemexpress LLC Dbco-peg4-acetic-val-cit-pab | 98.2% | 855.97 | C45H57N7O10 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY136098100MG
Encompass_Preferred
Cleavable 4-unit PEG antibody-drug conjugate (ADC) linker that contains a DBCO functional group for strain-promoted alkyne-azide cycloaddition (SPAAC). It is used to conjugate payloads to azide-containing molecules in ADC and bioconjugation workflows.
  • Cleavable Val-Cit-PAB linker for cathepsin B-mediated payload release.
  • DBCO moiety enables copper-free SPAAC with azide-functionalized partners.
  • PEG4 spacer improves solubility and reduces steric hindrance.
  • Available in multiple pack sizes, including 100 MG for synthesis scale-up.
  • High reported purity suitable for synthetic bioconjugation applications.
Catalog No. 50-002-03796