Learn More
Medchemexpress LLC DBCO-PEG4-C2-acid | 1537170-85-6 | 99.2% | 25 MG

Supplier: Medchemexpress LLC 25MGHY130396
DBCO-PEG4-C2-acid is a PEG-based PROTAC linker designed for the synthesis of PROTACs. It functions as a click chemistry reagent, featuring a DBCO group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with azide-containing molecules. This product appears as a white to yellow solid.
- PEG-based PROTAC linker
- Used in PROTACs synthesis
- Click chemistry reagent
- Contains a DBCO group
- Enables strain-promoted alkyne-azide cycloaddition (SPAAC)
- Appearance: White to yellow solid
- Molecular Weight: 552.62
- Formula: C30H36N2O8
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.