Learn More
Medchemexpress LLC Dbco-peg4-ggfg-dxd | 2694856-51-2 | 98.4% | 1375.45 | C72H79FN10O17 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY13472325MG
Encompass_Preferred
Dbco-peg4-ggfg-dxd is a click-reactive drug-linker conjugate for research use in antibody-drug conjugate (ADC) development. It contains a DBCO group for strain-promoted alkyne-azide cycloaddition to enable conjugation to azide-functionalized antibodies or biomolecules, a PEG4 spacer for solubility, a GGFG peptide linker, and a DXd topoisomerase I inhibitor payload.
  • Contains a DBCO click-reactive group for SPAAC conjugation.
  • Includes a PEG4 spacer to improve solubility and flexibility.
  • Features a GGFG peptide linker designed for cleavable release.
  • Delivers the DXd topoisomerase I inhibitor payload for cytotoxic activity.
  • Provided in small-milligram quantities suitable for research and ADC construction.
Catalog No. 50-002-03337