Learn More
Medchemexpress LLC Dbco-peg4-mmaf | 2360411-65-8 | 99.8% | 1266.56 g·mol⁻¹ | C69H99N7O15 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1334925MG
Encompass_Preferred
DBCO-PEG4-MMAF is a drug-linker conjugate used in antibody-drug conjugate research. It contains a dibenzocyclooctyne (DBCO) reactive group connected via a four-unit polyethylene glycol (PEG4) linker to the tubulin inhibitor MMAF, enabling strain-promoted alkyne-azide cycloaddition (SPAAC) for bioconjugation.
  • Contains DBCO reactive group for SPAAC click chemistry.
  • PEG4 spacer provides flexibility and improved solubility.
  • Includes MMAF payload, a potent tubulin polymerization inhibitor.
  • High purity and provided as a solid, white to off-white powder.
  • Available in small research pack sizes suitable for conjugation workflows.
Catalog No. 50-002-02410