Learn More
Medchemexpress LLC DBCO-PEG4-NHS ester | 1427004-19-0 | 99.0% | C34H39N3O10 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1402721G
Encompass_Preferred
DBCO-PEG4-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, containing a DBCO group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules. This product is intended for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains a DBCO group
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Reacts with azide-containing molecules
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins
  • For research use only
Catalog No. 50-003-57120