Learn More
Medchemexpress LLC DBCO-PEG4-NHS ester | 1427004-19-0 | 99.0% | C34H39N3O10 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY140272
Encompass_Preferred
DBCO-PEG4-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, containing a DBCO group capable of undergoing strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules that possess Azide groups.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Contains a DBCO group for SPAAC with Azide groups
  • For research use only
Catalog No. 50-003-52676