Learn More
Medchemexpress LLC DBCO-PEG4-NHS ester | 1427004-19-0 | 99.0% | C34H39N3O10 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14027250MG
Encompass_Preferred
DBCO-PEG4-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring a DBCO group capable of undergoing strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing azide groups. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • DBCO group enables strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Reacts with azide-containing molecules
  • PROTACs selectively degrade target proteins
Catalog No. 50-003-53034