Learn More
Medchemexpress LLC DBCO-PEG4-NHS ester | 1427004-19-0 | 99.0% | C34H39N3O10 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY140272500MG
Encompass_Preferred
DBCO-PEG4-NHS ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing a DBCO group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules possessing Azide groups. PROTACs leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains a DBCO group for strain-promoted alkyne-azide cycloaddition (SPAAC)
Catalog No. 50-003-57115