Learn More
Medchemexpress LLC Dbco-peg4-tamra | 1895849-41-8 | 96.2% | 936.06 | C54H57N5O10 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYD107010MG
Encompass_Preferred
DBCO-PEG4-TAMRA is a PEG-linked TAMRA fluorescent dye functionalized with a dibenzocyclooctyne (DBCO) group to enable copper-free strain-promoted alkyne-azide cycloaddition (SPAAC) for bioconjugation. It is supplied as a solid with high purity for labeling azide-containing biomolecules in imaging and analytical workflows.
  • Enables copper-free click chemistry conjugation.
  • PEG4 spacer improves solubility and reduces steric hindrance.
  • TAMRA fluorophore for red fluorescence detection.
  • Provided as a solid, light brown to brown in color.
  • Storage: store at -20°C, protect from light, under nitrogen.
  • High purity (≈96.2%) suitable for research applications.
Catalog No. 50-002-97175