Learn More
Medchemexpress LLC DBCO-PEG4-triethoxysilane | 2353410-02-1 | MFCD31811544 | 95.0% | C39H57N3O10Si | 10MG
SDP
Supplier:  Medchemexpress LLC HY14030810MG
Encompass_Preferred
DBCO-PEG4-triethoxysilane is a DBCO-functionalized PEG4 linker with a triethoxysilane end group, formulated for copper-free click chemistry and silane-based surface coupling. It is supplied as a colorless to light yellow liquid with good solubility in DMSO and is commonly provided at ~95% purity.
  • Provides a dibenzocyclooctyne (DBCO) group for copper-free click reactions.
  • Includes a PEG4 spacer to improve solubility and reduce steric hindrance.
  • Features a triethoxysilane moiety for covalent attachment to silica and glass surfaces.
  • Liquid form enables straightforward solution-phase handling and surface functionalization.
  • Soluble in common organic solvents such as DMSO, facilitating formulation and reactions.
Catalog No. 50-002-60364