Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dbco-PEG4-VA-PBD | 2241644-09-5 | 99.7% | 1430.60 | C80H87N9O16 | 1 MG
SDP

Supplier:  Medchemexpress LLC HY1334331MG

Encompass_Preferred

DBCO-PEG4-VA-PBD is a drug-linker conjugate intended for antibody-drug conjugate (ADC) conjugation workflows. The molecule features a DBCO reactive group for strain-promoted alkyne-azide cycloaddition (SPAAC), a PEG4 spacer, a valine-alanine protease-cleavable linker, and a pyrrolobenzodiazepine (PBD) cytotoxic payload. Supplied as a solid for research use.

  • Contains DBCO group for SPAAC click chemistry
  • Includes PEG4 spacer for improved solubility
  • Valine-alanine cleavable linker enables intracellular payload release
  • PBD payload for potent cytotoxic activity
  • Supplied as 1 mg solid; purity reported ~99.7%
  • Store at -80°C under nitrogen; shipped on dry ice

Catalog No. 50-002-01989


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.