Learn More
Medchemexpress LLC Dbco-PEG4-VA-PBD | 2241644-09-5 | MFCD00012576 | 99.7% | 1430.60 g·mol⁻1 | C80H87N9O16 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1334335MG
Encompass_Preferred
DBCO-PEG4-VA-PBD is a click chemistry reagent and drug-linker conjugate used to attach pyrrolobenzodiazepine (PBD) payloads to biomolecules via strain-promoted alkyne-azide cycloaddition (SPAAC). It contains a DBCO moiety, a PEG4 spacer, and a valine-alanine cleavable linker designed for antibody-drug conjugate (ADC) assembly.
  • Functions as a SPAAC-compatible click chemistry reagent.
  • Contains a PEG4 spacer and a valine-alanine cleavable linker.
  • Includes a pyrrolobenzodiazepine (PBD) cytotoxic payload.
  • High purity suitable for research applications.
  • Contains a single defined molecular formula and molecular weight for characterization.
  • Intended for research use in bioconjugation and ADC development.
Catalog No. 50-002-03317