Learn More
Medchemexpress LLC Dbco-PEG4-Val-Cit-PAB-MMAF | 2244602-23-9 | 98.7% | 25 MG
SDP
Supplier:  Medchemexpress LLC HY13099025MG
Encompass_Preferred
DBCO-PEG4-Val-Cit-PAB-MMAF is a drug-linker conjugate used in antibody-drug conjugate research. It combines a cleavable PEG-based Val-Cit-PAB linker bearing a DBCO moiety with the tubulin inhibitor MMAF to enable enzyme-triggered payload release and bioorthogonal conjugation via SPAAC.
  • Cleavable Val-Cit-PAB linker for cathepsin-mediated payload release.
  • DBCO moiety enables SPAAC click chemistry with azide-functionalized partners.
  • Contains MMAF tubulin inhibitor as the cytotoxic payload.
  • High purity suitable for research use (approximately 98.7%).
  • Available in small research pack sizes such as 25 mg.
Catalog No. 50-002-02391