Learn More
Medchemexpress LLC DBCO-PEG4-Val-Cit-PAB-PNP | 2226472-28-0 | 98.3% | 1079.16 g·mol⁻1 | C55H66N8O15 | 25 MG
SDP
Catalog No. 5000283162
Encompass_Preferred
DBCO-PEG4-Val-Cit-PAB-PNP is a cleavable antibody-drug conjugate (ADC) linker that integrates a dibenzocyclooctyne (DBCO) click handle, a four-unit PEG spacer, and a Val-Cit protease-cleavable dipeptide with a PAB-PNP leaving group. It is designed for strain-promoted azide-alkyne conjugation and for intracellular payload release following enzymatic cleavage, supporting ADC synthesis and other bioconjugation applications.
  • Enables copper-free click conjugation to azide-containing molecules.
  • Four-unit PEG spacer improves solubility and reduces steric hindrance.
  • Val-Cit dipeptide is cleavable by Cathepsin B for intracellular payload release.
  • PAB-PNP leaving group facilitates efficient payload attachment and release.
  • High purity suitable for synthetic and analytical workflows.
  • Stable storage and handling conditions support reproducible conjugation.
Catalog No. 50-002-83162 Supplier Medchemexpress LLC Supplier No. HYW19091325MG
DBCO-PEG4-Val-Cit-PAB-PNP is a cleavable antibody-drug conjugate (ADC) linker that integrates a dibenzocyclooctyne (DBCO) click handle, a four-unit PEG spacer, and a Val-Cit protease-cleavable dipeptide with a PAB-PNP leaving group. It is designed for strain-promoted azide-alkyne conjugation and for intracellular payload release following enzymatic cleavage, supporting ADC synthesis and other bioconjugation applications.
  • Enables copper-free click conjugation to azide-containing molecules.
  • Four-unit PEG spacer improves solubility and reduces steric hindrance.
  • Val-Cit dipeptide is cleavable by Cathepsin B for intracellular payload release.
  • PAB-PNP leaving group facilitates efficient payload attachment and release.
  • High purity suitable for synthetic and analytical workflows.
  • Stable storage and handling conditions support reproducible conjugation.