Learn More
Medchemexpress LLC Dbco-peg5-dbco | 2363130-04-3 | MFCD28334545 | 98.0% | 854.99 g/mol | C50H54N4O9 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1403025MG
Encompass_Preferred
DBCO-PEG5-DBCO is a bifunctional polyethylene glycol (PEG) linker bearing two dibenzocyclooctyne (DBCO) groups for copper-free click conjugation to azide-functionalized molecules. Supplied as a dry reagent for research-scale bioconjugation, PROTAC assembly, and probe development, it offers defined spacer length and high purity to support reliable coupling performance.
  • Enables copper-free strain-promoted azide-alkyne cycloaddition.
  • Contains two DBCO reactive termini for bifunctional linking.
  • PEG5 spacer reduces steric hindrance and improves solubility.
  • High purity supports reproducible bioconjugation.
  • Supplied in small-quantity packs for research-scale synthesis.
Catalog No. 50-002-22826