Learn More
Medchemexpress LLC DBCO-Val-Cit-PABC-PNP | 2994332-18-0 | 859.92 | C46H49N7O10 | 10 MG

Supplier: Medchemexpress LLC HY13093710MG
DBCO-Val-Cit-PABC-PNP is a cleavable antibody-drug conjugate (ADC) linker containing a DBCO group for strain-promoted alkyne-azide cycloaddition (SPAAC) and a Val-Cit-PABC protease-cleavable spacer. It is supplied as a solid reagent for research use in ADC synthesis.
- Contains DBCO for copper-free SPAAC click chemistry.
- Protease-cleavable Val-Cit-PABC spacer for enzyme-triggered payload release.
- Molecular weight 859.92 and formula C46H49N7O10.
- Available in small pack sizes suitable for synthesis, including 10 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.