Learn More
Medchemexpress LLC DBCO-Val-Cit-PABC-PNP | 2994332-18-0 | 97.7% | 859.92 | C46H49N7O10 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY130937100MG
Encompass_Preferred
DBCO-Val-Cit-PABC-PNP is a cleavable antibody-drug conjugate (ADC) linker that contains a DBCO clickable group, a Val-Cit peptide cleavage site, and a PABC self-immolative spacer. It is designed for click chemistry conjugation and controlled payload release in ADC synthesis.
  • Cleavable linker designed for cathepsin-mediated payload release.
  • DBCO clickable group enables strain-promoted azide-alkyne cycloaddition (SPAAC).
  • Val-Cit peptide motif provides a cleavable site for intracellular release.
  • PABC spacer facilitates self-immolative release of the payload.
  • High purity solid suitable for synthesis and conjugation workflows.
Catalog No. 50-002-03392