Learn More
Medchemexpress LLC DBCO-Val-Cit-PABC-PNP | 2994332-18-0 | 97.7% | 859.92 | C46H49N7O10 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13093750MG
Encompass_Preferred
DBCO-Val-Cit-PABC-PNP is a cleavable antibody-drug conjugate (ADC) linker that contains a dibenzocyclooctyne (DBCO) group for strain-promoted alkyne-azide cycloaddition (SPAAC). It is supplied as an off-white to light-yellow solid, with a molecular weight of 859.92, and is intended for use in ADC synthesis and click-chemistry conjugation workflows.
  • Cleavable val-cit dipeptide linker for enzymatic release in lysosomes.
  • DBCO group enables copper-free click conjugation (SPAAC) with azide-containing partners.
  • High purity suitable for research applications.
  • Available in small-scale package sizes for synthesis workflows.
  • Solid, off-white to light-yellow appearance; store per certificate of analysis.
Catalog No. 50-002-03928