Learn More
Medchemexpress LLC DBCO-Val-Cit-PABC-PNP | 2994332-18-0 | 97.7% | 859.92 g·mol⁻1 | C46H49N7O10 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1309371MG
Encompass_Preferred
DBCO-Val-Cit-PABC-PNP is a cleavable antibody-drug conjugate (ADC) linker that contains a dibenzocyclooctyne (DBCO) moiety for strain-promoted alkyne-azide cycloaddition (SPAAC). It is used as a click-chemistry reagent to attach payloads to azide-containing molecules and as a Val-Cit-PABC cleavable peptide linker in ADC synthesis.
  • Cleavable Val-Cit-PABC peptide linker susceptible to enzyme cleavage.
  • DBCO group enables copper-free SPAAC conjugation to azide-containing payloads.
  • High reported purity suitable for synthesis and analytical workflows.
  • Available in small research pack sizes for conjugation experiments.
Catalog No. 50-002-02515