Learn More
Abcam DBeQ, p97 ATPase inhibitor, 50MG
Supplier:  Abcam AB14125450MG
Encompass_Preferred
MW 340.4 Da, Purity >99%. Selective, potent, reversible and ATP-competitive p97 ATPase inhibitor (IC₅₀ = 1.5 μM). Inhibits a number of cellular processes including ER-associated degradation, ubiquitin fusion degradation and autophagosome maturation. Decreases cancer cell growth and induces activation of caspases 3 and 7.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6756