Learn More
Enzo Life Sciences dBET1 (5mg). CAS: 1799711-21-9
SDP
Supplier:  Enzo Life Sciences ENZCHM2600005
Encompass_Preferred
dBET1 is a PROTAC, consisting of a BRD4-ligand JQ-1 linked to a cereblon E3 ligase ligand, targeting on BET bromodomain BRD4. Purity: ≥99%. Long Term Storage: -20°C.
Catalog No. 50-201-3718