Learn More
*OAKTON INSTRUMENTS Waterproof pH Electrode with ATC, Double-Junction, Epoxy-Body, Gel-Filled for model 150/450
Supplier:  *OAKTON INSTRUMENTS 3580872/DEL
Encompass_Preferred
Gel filled. 6-pin waterproof. Double junction, epoxy body, waterproof pH electrode with ATC.
Catalog No. NC1154953