Learn More
Electron Microscopy Sciences Conductive Double Sided Carbon Tape with Aluminum, 12 mm 20 MT
SDP
Supplier:  Electron Microscopy Sciences 7781712AL
Encompass_Preferred
Can be used to mount particles 15-20 microns. Aluminum base.
Catalog No. 50-366-268