Learn More
Electron Microscopy Sciences DBP (Dibutyl Phthalate) 225 ML
SDP
Supplier:  Electron Microscopy Sciences 13101
Encompass_Preferred
CAS #84-74-2 C₆H₄(COOC₄H₉)₂ F.W. 278.35
A plasticizer for epoxy resins.
Catalog No. 50-980-356