Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dby HY Peptide (608-622), mouse | 288855-20-9 | 99.7% | 1568.56 | 1 MG
SDP

Supplier:  Medchemexpress LLC HYP53941MG

Encompass_Preferred

Dby HY Peptide (608-622), mouse, is a biologically active peptide. It is a HYAb epitope that belongs to a well-conserved family of genes coding for known or putative RNA helicases. This peptide contains a core sequence with a DEAD (Asp-Glu-Ala-Asp) box peptide motif, which is why it is named Dby (Dead box RNA helicase Y). The single Phenylalanine in its sequence acts as an anchor point, with FNSNRANSS likely being the core sequence of this HYAb epitope.

  • Biological active peptide
  • For research use only
  • High purity: 99.7%
  • Sealed storage, away from moisture
  • Powder storage: -80°C for 2 years or -20°C for 1 year
  • In solvent storage: -80°C for 6 months or -20°C for 1 month

Catalog No. 50-003-99136


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.