Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dby HY Peptide (608-622), mouse | 288855-20-9 | 99.7% | 1568.56 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYP539410MG

Encompass_Preferred

Dby HY Peptide (608-622), mouse is a biologically active peptide. It is a HYAb epitope belonging to a well-conserved family of genes coding for known or putative RNA helicases, containing a core sequence with a DEAD (Asp-Glu-Ala-Asp) box peptide motif. The single Phenylalanine in the sequence serves as the anchor point, and FNSNRANSS is most likely the "core" sequence of this HYAb epitope.

  • Biologically active peptide
  • HYAb epitope
  • Belongs to a family of genes coding for known or putative RNA helicases
  • Contains a core sequence with a DEAD (Asp-Glu-Ala-Asp) box peptide motif
  • Single Phenylalanine serves as an anchor point
  • FNSNRANSS is the core sequence of the HYAb epitope

Catalog No. 50-003-98930


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.