Learn More
Medchemexpress LLC DC-BPi-03 | 2758411-46-8 | C14H14N4O2S | 200 MG
SDP
Supplier:  Medchemexpress LLC HY150211200MG
Encompass_Preferred
DC-BPi-03 is a potent BPTF-BRD inhibitor with an IC50 of 698.3 nM and a Kd of 2.81 μM. This compound is intended for research use only.
  • Potent BPTF-BRD inhibitor
  • IC50 of 698.3 nM
  • Kd of 2.81 μM
  • Purity of 99.77%
  • White to off-white solid appearance
Catalog No. 50-003-75693