Learn More
Medchemexpress LLC DC-BPi-03 | 2758411-46-8 | C14H14N4O2S | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY150211
Encompass_Preferred
DC-BPi-03 is a potent BPTF-BRD inhibitor with an IC50 of 698.3 nM and a Kd of 2.81 μM. It is intended for research use only and is not for sale to patients.
  • Potent BPTF-BRD inhibitor
  • Inhibits BPTF-BRD with an IC50 of 698.3 nM
  • Inhibits BPTF-BRD with a Kd of 2.81 μM
  • For research use only
  • Relevant to cancer research
  • Relevant to blood or cardio-cerebrovascular disease research
  • Relevant to blood disease research
  • Relevant to leukemia/lymphoma/myeloma research
  • Relevant to cancer targeted therapy
  • Relevant to cancer metabolism and metastasis
  • Acts as an epigenetic reader domain inhibitor
Catalog No. 50-003-74506