Learn More
Medchemexpress LLC DC-BPi-11 (hydrochloride) | 97.0% | C20H24ClN5O2S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY141703A100MG
Encompass_Preferred
DC-BPi-11 hydrochloride is an inhibitor of bromodomain PHD finger transcription factor (BPTF), showing an IC50 value of 698 nM. It demonstrates significant inhibition against leukemia cell proliferation in human leukemia MV-4-11 cells with an EC50 value of 120 nM, while having minimal effects on normal cells, suggesting a favorable safety profile.
  • Inhibits bromodomain PHD finger transcription factor (BPTF)
  • Significantly inhibits leukemia cell proliferation
  • Decreases downstream oncogene expression
  • Dose-dependently decreases c-Myc protein level
  • Minimal effects on normal cells
Catalog No. 50-003-53405