Learn More
MOUSER ELECTRONICS DC FAN, 80X80X15MM, 12VDC, 37C
Supplier:  MOUSER ELECTRONICS ME80151V1000UA99
Encompass
DC Fan, 80x80x15mm, 12VDC, 37CFM, 1.96W, 34.7dBA, 3000RPM, Vapo, Auto Restart
Catalog No. NC2223252