Learn More
Bon Opus Biosciences Recombinan DC-SIGNR/CD299 (C-His) 50ug
SDP
Supplier:  Bon Opus Biosciences BP111350UG
Encompass_Preferred
Recombinant DC-SIGNR/CD299 (C-His) is produced by Human Cells. The target gene encoding Ser78-Glu399 is expressed with a C-His tag. Accession# Q9H2X3. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1113
Catalog No. 50-217-2709