Learn More
Medchemexpress LLC DC-U4106 | 2410534-62-0 | C29H27N5O5 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1505051MG
Encompass_Preferred
DC-U4106 is a USP8 targeting inhibitor with a Kd value of 4.7 μM and an IC50 value of 1.2 μM. It targets the ubiquitin pathway, facilitating the degradation of Erα. This compound inhibits tumor growth with minimal toxicity and shows potential for breast cancer research.
  • Targets the ubiquitin pathway
  • Facilitates degradation of Erα
  • Inhibits tumor growth with minimal toxicity
  • Reduces mRNA levels of ERα and PR
  • Regulates RTK pathway related proteins and ERα and PR protein expression
  • Induces apoptosis and inhibits cell proliferation
Catalog No. 50-003-75799