Learn More
Medchemexpress LLC HY-129379 1mg Medchemexpress, DC0-NH2 CAS:615538-51-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1293791MG
Encompass_Preferred
Medchemexpress, HY-129379 1mg DC0-NH2 CAS:615538-51-7 DC0-NH2 is an effector moiety for ADC and a simplified analog of DC1 with better stability. DC0-NH2 is about 1000-fold more cytotoxic than commonly used anticancer drugs (ex. Doxorubicin). DC0-NH2 can bind to the minor groove of DNA, followed by alkylation of adenine residues by its propabenzindole (CBI) component. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9805