Learn More
Medchemexpress LLC DC0-NH2 | 615538-51-7 | 95.0% | 550.01 | C31H24ClN5O3 | 10 MG

Supplier: Medchemexpress LLC HY12937910MG
DC0-NH2 is a small-molecule cytotoxin used as an effector moiety for antibody-drug conjugates (ADCs). It is a simplified analog of DC1 with improved stability and substantially higher cytotoxicity than several common anticancer agents, intended for research use as an ADC payload.
- Effector moiety for antibody-drug conjugates.
- Simplified analog of DC1 with improved stability.
- Approximately 1000-fold higher cytotoxicity than several common anticancer agents.
- Available as solid in multiple mg sizes and as a 10 mM solution in DMSO.
- Purity approximately 95.0%.
- Molecular weight 550.01 g/mol; formula C31H24ClN5O3.
- Appearance: light brown solid.
- Recommended storage: store under nitrogen at -20°C; in solvent store at -80°C up to 6 months.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.