Learn More
Medchemexpress LLC Dc07090 dihydrochloride | 1158264-37-9 | MFCD07612226 | 99.1% | 375.25 g·mol⁻¹ | C18H16Cl2N4O | 5MG

Supplier: Medchemexpress LLC HY1235175MG
DC07090 dihydrochloride is the dihydrochloride salt of a non-peptidyl inhibitor of human enterovirus 71 (EV71) 3C protease. It is low in toxicity, reversible, and competitive, with reported biochemical activity (IC50 21.72 μM; Ki 23.29 μM) and antiviral activity against coxsackievirus A16 (EC50 27.76 μM). Supplied as a light yellow solid for research use; recommended stock is 10 mM in DMSO.
- Potent, reversible, competitive EV71 3C protease inhibitor (IC50 21.72 μM).
- Demonstrated antiviral activity against coxsackievirus A16 (EC50 27.76 μM).
- High reported purity (99.12%).
- Good solubility in DMSO (100 mg/mL); in vivo formulation examples provided.
- Supplied as a light yellow solid; store sealed at 4°C, away from moisture.
- Recommended stock preparation: 10 mM in DMSO; in-solution stability: -80°C (6 months), -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.