Learn More
TARGETMOL CHEMICALS INC DC661 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T55385MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DC661 is palmitoyl-protein thioesterase 1 (PPT1) inhibitor inhibits autophagy and acts as an anti-lysosomal agent. Anti-cancer activitypurity: 98%
Catalog No. 50-257-9747