Learn More
Selleck Chemical LLC DC661 S8808-1G
SDP
Supplier:  Selleck Chemical LLC S88081G
Encompass_Preferred
DC661 is capable of deacidifying the lysosome and inhibiting autophagy significantly better than HCQ DC661 induces apoptosis
Catalog No. 50-000-24443