Learn More
TARGETMOL CHEMICALS INC DCBP-1 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T93705MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. dCBP-1 is a chemical degrader of p300/CBP. dCBP-1 hijacks the E3 ubiquitin ligase CRBN for selective degradation of p300/CBP. Degradation of p300/CBP by dCBP-1 leads to effective multiple myeloma cell killing. purity: 98%
Catalog No. 50-257-9007